Assay Details
Target Gene Details
Entrez Gene ID: | 9744 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 |
Gene Aliases: |
CENTB1 |
Location: |
Chr.17:7336506-7354944 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAP1 |
Target Gene Details
Entrez Gene ID: | 147040 |
Gene Name: | potassium channel tetramerization domain containing 11 |
Gene Aliases: |
C17orf36, KCASH1, REN, REN/KCTD11 |
Location: |
Chr.17:7351889-7354944 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD11 | NM_001002914.2 | 1 | 1176 | NP_001002914.1 |
| AK056227.1 | 1 | 903 | ||
| AY646650.1 | 1 | 903 | AAT75307.1 | |
| BC019929.1 | 1 | 427 | ||
| BC110598.2 | 1 | 531 | AAI10599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv952118 | Chr.17:7174582 - 7456981 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map