Assay Details
Target Gene Details
Entrez Gene ID: | 2247 |
Gene Name: | fibroblast growth factor 2 |
Gene Aliases: |
BFGF, FGF-2, FGFB, HBGF-2 |
Location: |
Chr.4:122826708-122898235 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF2 | NM_002006.4 | NP_001997.5 | ||
| AB451321.1 | ||||
| AB451450.1 | ||||
| AY820133.1 | AAV70487.1 | |||
| J04513.1 | AAA52532.1 | |||
| M17599.1 | AAA52534.1 | |||
| M27968.1 | AAA52448.1 | |||
| S47380.1 | AAD13853.1 |
More Information
Set Membership: |
|

Genomic Map