Assay Details
Target Gene Details
Entrez Gene ID: | 6296 |
Gene Name: | acyl-CoA synthetase medium-chain family member 3 |
Gene Aliases: |
SA, SAH |
Location: |
Chr.16:20763990-20797157 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSM3 | NM_005622.3 | 10 | 1450 | NP_005613.2 |
| XM_011545911.1 | 11 | 1555 | XP_011544213.1 | |
| XM_017023523.1 | 10 | 1444 | XP_016879012.1 | |
| XM_017023524.1 | 9 | 1314 | XP_016879013.1 | |
| AK223139.1 | 10 | 1419 | BAD96859.1 | |
| AK290322.1 | 10 | 1419 | ||
| AK296122.1 | 10 | 1249 | ||
| BI826935.1 | 4 | 325 | ||
| BU170035.1 | 4 | 302 | ||
| D16350.1 | 9 | 1290 | BAA03853.1 |
Target Gene Details
Entrez Gene ID: | 112479 |
Gene Name: | ERI1 exoribonuclease family member 2 |
Gene Aliases: |
EXOD1, ZGRF5 |
Location: |
Chr.16:20780193-20806473 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERI2 | NM_080663.2 | NP_542394.2 | ||
| BC010503.1 | AAH10503.1 | |||
| BX437401.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050009 | Chr.16:20773077 - 20862801 on Build GRCh38 | Loss |
|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|
| nsv833165 | Chr.16:20718218 - 20886396 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map