Assay Details
Target Gene Details
Entrez Gene ID: | 372 |
Gene Name: | archain 1 |
Gene Aliases: |
COPD |
Location: |
Chr.11:118572363-118603033 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARCN1 | NM_001142281.1 | NP_001135753.1 | ||
| NM_001655.4 | NP_001646.2 | |||
| XM_005271542.3 | XP_005271599.1 | |||
| AK304019.1 | ||||
| AK312298.1 | ||||
| BC064936.1 | AAH64936.1 | |||
| BC093636.1 | AAH93636.1 | |||
| BC093638.1 | AAH93638.1 | |||
| BI560739.1 | ||||
| BX640860.1 | CAE45922.1 | |||
| DB216489.1 | ||||
| X81197.1 | CAA57071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78730658] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map