Assay Details
Target Gene Details
Entrez Gene ID: | 79682 |
Gene Name: | centromere protein U |
Gene Aliases: |
CENP50, CENPU50, KLIP1, MLF1IP, PBIP1 |
Location: |
Chr.4:184694065-184734132 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPU | NM_024629.3 | 13 | 2081 | NP_078905.2 |
| NR_104593.1 | 12 | 2019 | ||
| XM_005263218.4 | 13 | 2116 | XP_005263275.2 | |
| AF516710.1 | 14 | 2498 | AAQ08228.1 | |
| AW440254.1 | 1 | 474 | ||
| BX355581.2 | 1 | 416 |
Target Gene Details
Entrez Gene ID: | 201973 |
Gene Name: | primase and DNA directed polymerase |
Gene Aliases: |
CCDC111, MYP22 |
Location: |
Chr.4:184649613-184694959 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss |
|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145762735] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map