Assay Details
Target Gene Details
Entrez Gene ID: | 55719 |
Gene Name: | SMC5-SMC6 complex localization factor 2 |
Gene Aliases: |
C10orf6, FAM178A |
Location: |
Chr.10:100910230-100965134 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLF2 | NM_001136123.1 | 1 | 510 | NP_001129595.1 |
| NM_001243770.1 | 1 | 510 | NP_001230699.1 | |
| NM_018121.3 | 1 | 510 | NP_060591.3 | |
| XM_005269965.2 | 1 | 128 | XP_005270022.1 | |
| XM_011539944.2 | 1 | 2849 | XP_011538246.1 | |
| AF460991.1 | 1 | 510 | AAN84475.1 | |
| AF460992.1 | 1 | 510 | ||
| AK292565.1 | 1 | 127 | ||
| AL832343.1 | 1 | 116 | ||
| AL832383.1 | 1 | 78 | ||
| AL833126.1 | 1 | 117 | ||
| BC020915.1 | 1 | 92 | AAH20915.1 | |
| BC047715.1 | 1 | 106 | ||
| BC073832.1 | 1 | 83 | AAH73832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv552051 | Chr.10:100904892 - 100984619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map