Assay Details
Target Gene Details
Entrez Gene ID: | 55650 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class V |
Gene Aliases: |
GPI-MT-II, HPMRS1, PIG-V |
Location: |
Chr.1:26787963-26798403 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGV | NM_001202554.1 | NP_001189483.1 | ||
| NM_017837.3 | 1 | 230 | NP_060307.2 | |
| AK000484.1 | BAA91196.1 | |||
| AK056539.1 | ||||
| AK225521.1 | ||||
| AK301626.1 | ||||
| BC013568.1 | 1 | 198 | AAH13568.1 | |
| BI463967.1 | ||||
| DA280125.1 | 1 | 230 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map