Assay Details
Target Gene Details
Entrez Gene ID: | 201633 |
Gene Name: | T-cell immunoreceptor with Ig and ITIM domains |
Gene Aliases: |
VSIG9, VSTM3, WUCAM |
Location: |
Chr.3:114293986-114310288 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIGIT | NM_173799.3 | 4 | 2124 | NP_776160.2 |
| XM_011512538.1 | 4 | 2109 | XP_011510840.1 | |
| XM_017005865.1 | 3 | 2399 | XP_016861354.1 | |
| AK097192.1 | BAC04973.1 | |||
| BX640915.1 | 3 | 2003 | CAE45956.1 |
More Information
Set Membership: |
|

Genomic Map