Assay Details
Target Gene Details
Entrez Gene ID: | 6414 |
Gene Name: | selenoprotein P, plasma, 1 |
Gene Aliases: |
SELP, SEPP, SeP |
Location: |
Chr.5:42799880-42811922 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPP1 | NM_001085486.1 | 3 | 255 | NP_001078955.1 |
| NM_001093726.1 | 3 | 314 | NP_001087195.1 | |
| NM_005410.2 | 2 | 226 | NP_005401.3 | |
| AK096125.1 | 2 | 595 | ||
| AK225801.1 | 3 | 225 | ||
| AK311392.1 | 2 | 563 | ||
| AL833145.1 | 2 | 197 | ||
| AU136519.2 | 3 | 284 | ||
| BC015875.1 | 2 | 200 | AAH15875.1 | |
| BC040075.1 | 2 | 176 | AAH40075.1 | |
| BC046152.1 | 2 | 177 | AAH46152.1 | |
| BC058919.1 | 2 | 207 | AAH58919.1 | |
| BI463468.1 | 2 | 371 | ||
| Z11793.1 | 2 | 162 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5637n100 | Chr.5:42609988 - 42844089 on Build GRCh38 | Gain |
|
| nsv597886 | Chr.5:42610307 - 42854306 on Build GRCh38 | Gain |
|
| nsv523033 | Chr.5:42751935 - 42919583 on Build GRCh38 | Gain |
|
| nsv462126 | Chr.5:42728729 - 43237086 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72554691] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map