Assay Details
Target Gene Details
Entrez Gene ID: | 10352 |
Gene Name: | tryptophanyl tRNA synthetase 2, mitochondrial |
Gene Aliases: |
TrpRS |
Location: |
Chr.1:119031216-119140701 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WARS2 | NM_015836.3 | 6 | 1731 | NP_056651.1 |
| NM_201263.2 | 6 | 1760 | NP_957715.1 | |
| XM_005270350.3 | 6 | 1817 | XP_005270407.1 | |
| XM_011540494.2 | 7 | 1800 | XP_011538796.1 | |
| XM_011540495.2 | 5 | 1502 | XP_011538797.1 | |
| XM_017000038.1 | 5 | 1666 | XP_016855527.1 | |
| XM_017000039.1 | 8 | 1986 | XP_016855528.1 | |
| XM_017000040.1 | 6 | 1553 | XP_016855529.1 | |
| XM_017000041.1 | 5 | 1643 | XP_016855530.1 | |
| XM_017000042.1 | 5 | 1637 | XP_016855531.1 | |
| AK223197.1 | 6 | 1709 | BAD96917.1 | |
| BC044575.1 | 6 | 1714 | AAH44575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997707 | Chr.1:118656945 - 119053326 on Build GRCh38 | Gain |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74112264] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map