Assay Details
Target Gene Details
Entrez Gene ID: | 1828 |
Gene Name: | desmoglein 1 |
Gene Aliases: |
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1 |
Location: |
Chr.18:31318089-31359246 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1 | NM_001942.3 | 15 | 4285 | NP_001933.2 |
| AF088042.1 | 1 | 377 | ||
| AF097935.1 | 15 | 4286 | AAC83817.1 |
Target Gene Details
Entrez Gene ID: | 101927718 |
Gene Name: | DSG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:31343368-31426988 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSG1-AS1 | NR_110788.1 | |||
| BX476374.1 | ||||
| BX476414.1 |
More Information
Set Membership: |
|

Genomic Map