Assay Details
Target Gene Details
Entrez Gene ID: | 2686 |
Gene Name: | gamma-glutamyltransferase 7 |
Gene Aliases: |
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2 |
Location: |
Chr.20:34844720-34872858 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGT7 | NM_178026.2 | NP_821158.2 | ||
| XM_011528782.2 | XP_011527084.1 | |||
| XM_011528783.2 | XP_011527085.1 | |||
| XM_017027809.1 | XP_016883298.1 | |||
| XM_017027810.1 | XP_016883299.1 | |||
| AK090413.1 | 5 | 5701 | BAC03394.1 | |
| BC033745.1 | ||||
| BC101370.1 | ||||
| BC101371.1 | ||||
| BC101372.1 | ||||
| BC141843.1 | ||||
| DA117929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
| esv3567955 | Chr.20:34838184 - 34853908 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map