Assay Details
Target Gene Details
Entrez Gene ID: | 102546294 |
Gene Name: | uncharacterized LOC102546294 |
Gene Aliases: |
- |
Location: |
Chr.5:148268307-148383850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102546294 | NR_105057.1 | |||
| AK054753.1 | ||||
| DA523579.1 |
Target Gene Details
Entrez Gene ID: | 643394 |
Gene Name: | serine peptidase inhibitor, Kazal type 9 |
Gene Aliases: |
LEKTI2 |
Location: |
Chr.5:148323219-148341974 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK9 | NM_001040433.1 | NP_001035523.1 | ||
| XM_017009709.1 | XP_016865198.1 | |||
| AY396740.1 | AAR91618.1 | |||
| BC137530.1 | ||||
| BC137531.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss |
|
| esv3607106 | Chr.5:148224310 - 148363295 on Build GRCh38 | Loss |
|
| nsv5058 | Chr.5:148313602 - 148346355 on Build GRCh38 | Insertion |
|
| nsv599945 | Chr.5:148269618 - 148340924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map