Assay Details
Target Gene Details
Entrez Gene ID: | 112495 |
Gene Name: | general transcription factor IIIC subunit 6 |
Gene Aliases: |
C6orf51, TFIIIC35, bA397G5.3 |
Location: |
Chr.6:110958560-110967888 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C6 | NM_138408.3 | NP_612417.1 | ||
| AF361492.1 | AAL57217.1 | |||
| AF361496.1 | AAL57221.1 | |||
| AK057977.1 | BAB71624.1 | |||
| BC011593.1 | AAH11593.1 | |||
| BC104655.1 | AAI04656.1 | |||
| BQ269752.1 | ||||
| EF137904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain |
|
| nsv1016001 | Chr.6:110959208 - 110988298 on Build GRCh38 | Gain |
|
| nsv830773 | Chr.6:110813116 - 110985229 on Build GRCh38 | Loss |
|
| nsv509148 | Chr.6:110947375 - 110967436 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map