Assay Details
Target Gene Details
Entrez Gene ID: | 4343 |
Gene Name: | Mov10 RISC complex RNA helicase |
Gene Aliases: |
fSAP113, gb110 |
Location: |
Chr.1:112674312-112702377 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOV10 | NM_001130079.2 | NP_001123551.1 | ||
| NM_001286072.1 | NP_001273001.1 | |||
| NM_001321324.1 | NP_001308253.1 | |||
| NM_020963.4 | NP_066014.1 | |||
| XM_005270869.4 | XP_005270926.1 | |||
| XM_005270870.4 | XP_005270927.1 | |||
| XM_017001318.1 | XP_016856807.1 | |||
| XM_017001319.1 | XP_016856808.1 | |||
| AB046851.1 | ||||
| AK023297.1 | BAB14514.1 | |||
| AK074174.1 | BAB85000.1 | |||
| AK226011.1 | ||||
| AK301229.1 | ||||
| AK308874.1 | ||||
| AK310512.1 | ||||
| AL833353.1 | ||||
| BC002548.1 | AAH02548.1 | |||
| BC004499.2 | AAH04499.2 | |||
| BC009312.2 | AAH09312.1 | |||
| BX647787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv997362 | Chr.1:112692198 - 112700675 on Build GRCh38 | Deletion |
|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map