Assay Details
Target Gene Details
Entrez Gene ID: | 284361 |
Gene Name: | ER membrane protein complex subunit 10 |
Gene Aliases: |
C19orf63, HSM1, HSS1 |
Location: |
Chr.19:50476477-50505905 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC10 | NM_175063.5 | NP_778233.4 | ||
| NM_206538.3 | NP_996261.1 | |||
| XM_006723163.3 | XP_006723226.1 | |||
| XM_011526818.2 | XP_011525120.1 | |||
| AY194293.2 | ||||
| AY358710.1 | AAQ89073.1 | |||
| AY761095.1 | AAV30543.1 | |||
| AY761097.1 | AAV30545.1 | |||
| BC032948.2 | AAH32948.1 | |||
| BC035001.2 | AAH35001.1 | |||
| BX366174.2 | ||||
| HY107516.1 |
Target Gene Details
Entrez Gene ID: | 112703 |
Gene Name: | family with sequence similarity 71 member E1 |
Gene Aliases: |
- |
Location: |
Chr.19:50466643-50476746 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71E1 | NM_001308429.1 | NP_001295358.1 | ||
| NM_138411.2 | NP_612420.1 | |||
| BC071737.1 | AAH71737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map