Assay Details
Target Gene Details
Entrez Gene ID: | 6399 |
Gene Name: | trafficking protein particle complex 2 |
Gene Aliases: |
MIP2A, SEDL, SEDT, TRAPPC2P1, TRS20, ZNF547L, hYP38334 |
Location: |
Chr.X:13712242-13734635 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC2 | NM_001011658.3 | NP_001011658.1 | ||
| NM_001128835.2 | NP_001122307.2 | |||
| NM_014563.5 | NP_055378.1 | |||
| XM_011545565.1 | XP_011543867.1 | |||
| XM_011545566.2 | XP_011543868.1 | |||
| AK310542.1 | ||||
| AK313736.1 | ||||
| BC016915.1 | AAH16915.1 | |||
| BC052618.1 | AAH52618.1 | |||
| BI821518.1 | ||||
| CB962763.1 | ||||
| CD698535.1 | ||||
| DA344866.1 | ||||
| DA350753.1 | ||||
| DA542477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528634 | Chr.X:13696631 - 14038859 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map