Assay Details
Target Gene Details
Entrez Gene ID: | 158248 |
Gene Name: | tetratricopeptide repeat domain 16 |
Gene Aliases: |
- |
Location: |
Chr.9:127716064-127731600 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC16 | NM_001317037.1 | 2 | 190 | NP_001303966.1 |
| NM_144965.2 | 2 | 190 | NP_659402.1 | |
| XM_005251733.1 | XP_005251790.1 | |||
| XM_005251734.1 | 2 | 175 | XP_005251791.1 | |
| XM_006716970.1 | 2 | 175 | XP_006717033.1 | |
| XM_006716971.1 | XP_006717034.1 | |||
| XM_011518283.1 | 2 | 188 | XP_011516585.1 | |
| XM_011518284.1 | 2 | 190 | XP_011516586.1 | |
| XM_011518285.1 | 2 | 188 | XP_011516587.1 | |
| XM_011518288.1 | 2 | 190 | XP_011516590.1 | |
| XM_011518289.2 | 2 | 138 | XP_011516591.1 | |
| XM_011518290.1 | 2 | 190 | XP_011516592.1 | |
| XM_011518291.1 | 2 | 175 | XP_011516593.1 | |
| XM_011518292.1 | 2 | 190 | XP_011516594.1 | |
| XM_011518293.1 | 2 | 188 | XP_011516595.1 | |
| XM_011518294.2 | 2 | 188 | XP_011516596.1 | |
| XM_011518295.1 | 2 | 188 | XP_011516597.1 | |
| XM_011518297.1 | 2 | 188 | XP_011516599.1 | |
| XM_011518298.2 | 2 | 188 | XP_011516600.1 | |
| AK057342.1 | 2 | 175 | BAB71437.1 | |
| AK294862.1 | 2 | 190 | ||
| AK302422.1 | ||||
| AK302737.1 | 2 | 175 | ||
| BC031281.1 | 2 | 188 | AAH31281.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|

Genomic Map