Assay Details
Target Gene Details
Entrez Gene ID: | 25960 |
Gene Name: | adhesion G protein-coupled receptor A2 |
Gene Aliases: |
GPR124, TEM5 |
Location: |
Chr.8:37796883-37843986 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRA2 | NM_032777.9 | NP_116166.9 | ||
| XM_005273471.3 | XP_005273528.1 | |||
| XM_011544481.2 | XP_011542783.1 | |||
| XM_011544482.2 | XP_011542784.1 | |||
| XM_011544483.2 | XP_011542785.1 | |||
| AB040964.2 | BAA96055.2 | |||
| AF378755.1 | AAL11992.1 | |||
| AK027296.1 | BAB55022.1 | |||
| AK074099.2 | ||||
| AY090637.1 | AAM09084.1 | |||
| AY181242.1 | AAO27354.1 | |||
| BC146774.1 | ||||
| DA569569.1 | ||||
| KU257672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509261 | Chr.8:37767151 - 37881434 on Build GRCh38 | Insertion |
|
| nsv528524 | Chr.8:37789148 - 37839356 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map