Assay Details
Target Gene Details
Entrez Gene ID: | 117246 |
Gene Name: | FtsJ homolog 3 |
Gene Aliases: |
EPCS3, SPB1 |
Location: |
Chr.17:63819433-63827671 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FTSJ3 | NM_017647.3 | 17 | 2565 | NP_060117.3 |
| AF327355.1 | 16 | 2040 | AAL56015.1 | |
| AK000069.1 | 9 | 1186 | BAA90924.1 | |
| AK095861.1 | 17 | 2565 | ||
| AK296715.1 | 16 | 1771 | ||
| AK307398.1 | 16 | 2054 | ||
| AK315010.1 | 17 | 1990 | ||
| AL834482.1 | 11 | 1453 | CAD39141.1 | |
| BC000131.1 | 12 | 1580 | AAH00131.2 | |
| BC036710.2 | 16 | 1949 | AAH36710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833510 | Chr.17:63767293 - 63927790 on Build GRCh38 | Loss |
|
| nsv517272 | Chr.17:63814307 - 64020132 on Build GRCh38 | Gain+Loss |
|
| nsv510720 | Chr.17:63809351 - 63908780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map