Assay Details
Target Gene Details
Entrez Gene ID: | 57184 |
Gene Name: | family with sequence similarity 219 member B |
Gene Aliases: |
C15orf17 |
Location: |
Chr.15:74898763-74907228 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM219B | NM_001321920.1 | NP_001308849.1 | ||
| NM_001321921.1 | NP_001308850.1 | |||
| NM_001321922.1 | NP_001308851.1 | |||
| NM_001321923.1 | NP_001308852.1 | |||
| NM_020447.4 | NP_065180.1 | |||
| NR_135841.1 | ||||
| NR_135842.1 | ||||
| NR_135843.1 | ||||
| NR_135844.1 | ||||
| NR_135845.1 | ||||
| NR_135846.1 | ||||
| NR_135847.1 | ||||
| NR_135848.1 | ||||
| NR_135849.1 | ||||
| NR_135850.1 | ||||
| NR_135851.1 | ||||
| NR_135852.1 | ||||
| NR_135853.1 | ||||
| NR_135854.1 | ||||
| XM_017022433.1 | XP_016877922.1 | |||
| AK000005.1 | BAA92230.1 | |||
| AK001572.1 | ||||
| AK291020.1 | ||||
| AK296403.1 | ||||
| AK308954.1 | ||||
| BC015637.2 | AAH15637.1 | |||
| DB153105.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819808 | Chr.15:74906071 - 74907183 on Build GRCh38 | Gain |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|

Genomic Map