Assay Details
Target Gene Details
Entrez Gene ID: | 5250 |
Gene Name: | solute carrier family 25 member 3 |
Gene Aliases: |
OK/SW-cl.48, PHC, PTP |
Location: |
Chr.12:98593625-98602000 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A3 | NM_002635.3 | 8 | 1521 | NP_002626.1 |
| NM_005888.3 | 8 | 1524 | NP_005879.1 | |
| NM_213611.2 | 7 | 1755 | NP_998776.1 | |
| BX647062.1 | 7 | 3077 | CAE46080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559918 | Chr.12:98570959 - 98666572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map