Assay Details
Target Gene Details
Entrez Gene ID: | 56339 |
Gene Name: | methyltransferase like 3 |
Gene Aliases: |
IME4, M6A, MT-A70, Spo8 |
Location: |
Chr.14:21498131-21511375 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL3 | NM_019852.4 | 10 | 1729 | NP_062826.2 |
| XM_006720206.3 | 10 | 1630 | XP_006720269.1 | |
| XM_011536968.1 | 9 | 1658 | XP_011535270.1 | |
| AK000222.1 | 10 | 1652 | ||
| AK093509.1 | 8 | 1511 | ||
| AK097873.1 | 4 | 2244 | ||
| AK098655.1 | 7 | 1171 | ||
| BC001650.2 | 10 | 1663 | AAH01650.1 | |
| BC003031.1 | 10 | 1663 | AAH03031.1 | |
| BC052244.1 | 10 | 1613 | AAH52244.1 | |
| BX247964.1 | 8 | 1458 | CAD62303.1 | |
| CA311761.1 | 2 | 310 |
Target Gene Details
Entrez Gene ID: | 9878 |
Gene Name: | TOX high mobility group box family member 4 |
Gene Aliases: |
C14orf92, KIAA0737, LCP1, MIG7 |
Location: |
Chr.14:21477176-21499177 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOX4 | NM_001303523.1 | 8 | 4373 | NP_001290452.1 |
| NM_014828.3 | 9 | 4442 | NP_055643.1 | |
| AY305872.1 | 1 | 2248 | AAQ95214.1 | |
| BU677026.1 | 1 | 111 | ||
| N63399.1 | 1 | 104 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633718 | Chr.14:21495207 - 21527525 on Build GRCh38 | Gain |
|
| esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
|
| nsv85879 | Chr.14:21497546 - 21506056 on Build GRCh38 | Deletion |
|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map