Assay Details
Target Gene Details
Entrez Gene ID: | 51202 |
Gene Name: | DEAD-box helicase 47 |
Gene Aliases: |
E4-DBP, HQ0256, MSTP162, RRP3 |
Location: |
Chr.12:12813346-12829981 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX47 | NM_016355.3 | NP_057439.2 | ||
| NM_201224.1 | NP_957518.1 | |||
| AF078843.1 | AAF23354.1 | |||
| AF190165.1 | AAQ13709.1 | |||
| AJ276704.1 | CAC14786.1 | |||
| AK054574.1 | BAB70762.1 | |||
| AK222940.1 | BAD96660.1 | |||
| AK225144.1 | ||||
| AK302536.1 | ||||
| AK310501.1 | ||||
| AL136666.1 | CAB66601.1 | |||
| AL833469.1 | ||||
| BC009379.2 | AAH09379.2 | |||
| BC068009.1 | AAH68009.1 | |||
| EF036507.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map