Assay Details
Target Gene Details
Entrez Gene ID: | 4297 |
Gene Name: | lysine methyltransferase 2A |
Gene Aliases: |
ALL-1, CXXC7, HRX, HTRX1, MLL, MLL-AF9, MLL/GAS7, MLL1, MLL1A, TET1-MLL, TRX1, WDSTS |
Location: |
Chr.11:118436490-118526832 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 35 - Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT2A | NM_001197104.1 | NP_001184033.1 | ||
| NM_005933.3 | NP_005924.2 | |||
| XM_006718839.3 | XP_006718902.2 | |||
| XM_011542829.2 | XP_011541131.1 | |||
| XM_011542830.2 | XP_011541132.1 | |||
| XM_011542831.2 | XP_011541133.1 | |||
| XM_011542833.2 | XP_011541135.1 | |||
| AB209508.1 | BAD92745.1 | |||
| AK295635.1 | ||||
| L04284.1 | AAA58669.1 | |||
| L04731.1 |
Target Gene Details
Entrez Gene ID: | 101929089 |
Gene Name: | uncharacterized LOC101929089 |
Gene Aliases: |
- |
Location: |
Chr.11:118511918-118531094 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929089 | NR_120572.1 | |||
| NR_120573.1 | ||||
| NR_120574.1 | ||||
| NR_120575.1 | ||||
| NR_120576.1 | ||||
| AI885123.1 | ||||
| AW449653.1 | ||||
| AW450539.1 | ||||
| AW611756.1 | ||||
| BE671460.1 | ||||
| BM930081.1 | ||||
| BU680885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
|
| esv3627872 | Chr.11:118502100 - 118536457 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map