Assay Details
Target Gene Details
Entrez Gene ID: | 754 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
C21orf1, C21orf3, PBF |
Location: |
Chr.21:44849585-44873903 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTTG1IP | NM_001286822.1 | 3 | 1594 | NP_001273751.1 |
| NM_004339.3 | 6 | 1922 | NP_004330.1 | |
| NR_104597.1 | 5 | 1813 | ||
| AF149785.1 | 6 | 1860 | AAF73770.1 | |
| AK094882.1 | 7 | 1627 | ||
| AK095586.1 | 1 | 2222 | ||
| BC000415.2 | 6 | 1679 | AAH00415.1 | |
| BC012858.1 | 6 | 1705 | AAH12858.1 | |
| BC019295.1 | 6 | 1669 | AAH19295.1 | |
| BC020983.1 | 6 | 1686 | AAH20983.1 | |
| BC031097.1 | 7 | 1650 | AAH31097.1 | |
| BC034250.1 | 6 | 1694 | AAH34250.1 | |
| L48984.1 | 3 | 1320 | ||
| Z50022.1 | 6 | 1728 | CAA90325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058550 | Chr.21:44807248 - 44898019 on Build GRCh38 | Loss |
|
| esv3647136 | Chr.21:44802580 - 44905193 on Build GRCh38 | Loss |
|
| nsv459298 | Chr.21:44771532 - 44858053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map