Assay Details
Target Gene Details
Entrez Gene ID: | 5144 |
Gene Name: | phosphodiesterase 4D |
Gene Aliases: |
ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1 |
Location: |
Chr.5:58969038-60488098 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830312 | Chr.5:58895752 - 59086399 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map