Assay Details
Target Gene Details
Entrez Gene ID: | 6547 |
Gene Name: | solute carrier family 8 member A3 |
Gene Aliases: |
NCX3 |
Location: |
Chr.14:70044217-70189626 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC8A3 | NM_001130417.2 | 6 | 2683 | NP_001123889.1 |
| NM_033262.4 | 8 | 4980 | NP_150287.1 | |
| NM_058240.3 | 8 | 4977 | NP_489479.1 | |
| NM_182932.2 | 7 | 4968 | NP_891977.1 | |
| NM_182936.2 | 5 | 2579 | NP_891981.1 | |
| NM_183002.2 | 8 | 4986 | NP_892114.1 | |
| NR_104122.1 | 6 | 4864 | ||
| XM_006720240.3 | 7 | 3832 | XP_006720303.1 | |
| XM_017021606.1 | 8 | 4358 | XP_016877095.1 | |
| XM_017021607.1 | 8 | 4848 | XP_016877096.1 | |
| XM_017021608.1 | 7 | 4594 | XP_016877097.1 | |
| XM_017021609.1 | 7 | 4830 | XP_016877098.1 | |
| XM_017021611.1 | 6 | 2956 | XP_016877100.1 | |
| AF510501.1 | 7 | 4968 | AAN60790.1 | |
| AF510502.1 | 8 | 4986 | AAN60791.1 | |
| AF510503.1 | 6 | 4864 | AAN60792.1 | |
| AJ508602.1 | 6 | 3764 | CAD48420.1 | |
| AJ745101.1 | 5 | 2542 | CAG33739.1 | |
| AJ745102.1 | 6 | 2646 | CAG33740.1 | |
| AK096553.1 | 5 | 2509 | ||
| AK122728.1 | 6 | 2670 | ||
| BC142969.1 | 6 | 2672 | ||
| BX098112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565007 | Chr.14:69615239 - 70107432 on Build GRCh38 | Gain |
|
| nsv832824 | Chr.14:69853922 - 70044684 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map