Assay Details
Target Gene Details
Entrez Gene ID: | 9146 |
Gene Name: | hepatocyte growth factor-regulated tyrosine kinase substrate |
Gene Aliases: |
HRS |
Location: |
Chr.17:81683932-81702121 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HGS | NM_004712.4 | NP_004703.1 | ||
| XM_011525463.2 | XP_011523765.1 | |||
| XM_017025297.1 | XP_016880786.1 | |||
| XM_017025298.1 | XP_016880787.1 | |||
| AF260566.1 | AAF82361.1 | |||
| AK294190.1 | ||||
| AK303800.1 | ||||
| BC003565.1 | AAH03565.1 | |||
| BT009754.1 | AAP88756.1 | |||
| D84064.1 | BAA23366.1 | |||
| DR001213.1 | ||||
| KU178466.1 | ||||
| KU178467.1 | ||||
| U43895.1 | AAC51929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss |
|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss |
|
| nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72858892,rs72858893] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map