Assay Details
Target Gene Details
Entrez Gene ID: | 2246 |
Gene Name: | fibroblast growth factor 1 |
Gene Aliases: |
AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1 |
Location: |
Chr.5:142592178-142698070 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF1 | NM_000800.4 | 4 | 1699 | NP_000791.1 |
| NM_001144892.2 | 4 | 1219 | NP_001138364.1 | |
| NM_001144934.1 | 5 | 1412 | NP_001138406.1 | |
| NM_001144935.1 | 4 | 1318 | NP_001138407.1 | |
| NM_001257205.1 | 4 | 1609 | NP_001244134.1 | |
| NM_001257206.1 | 4 | 1606 | NP_001244135.1 | |
| NM_001257207.1 | 5 | 1352 | NP_001244136.1 | |
| NM_001257208.1 | 4 | 1350 | NP_001244137.1 | |
| NM_001257209.1 | 4 | 1365 | NP_001244138.1 | |
| NM_001257210.1 | 4 | 1349 | NP_001244139.1 | |
| NM_001257211.1 | 4 | 1347 | NP_001244140.1 | |
| NM_001257212.1 | 4 | 1216 | NP_001244141.1 | |
| NM_033136.3 | 3 | 1595 | NP_149127.1 | |
| NM_033137.2 | 2 | 1053 | NP_149128.1 | |
| XM_005268390.4 | 4 | 1289 | XP_005268447.1 | |
| XM_011537607.2 | 5 | 1409 | XP_011535909.1 | |
| X51943.1 | 3 | 1160 | CAA36206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034911 | Chr.5:142554309 - 142652186 on Build GRCh38 | Gain |
|
| esv3606999 | Chr.5:142429551 - 143065624 on Build GRCh38 | Gain |
|
| nsv1031126 | Chr.5:142514198 - 143020006 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map