Assay Details
Target Gene Details
Entrez Gene ID: | 90627 |
Gene Name: | StAR related lipid transfer domain containing 13 |
Gene Aliases: |
ARHGAP37, DLC2, GT650, LINC00464 |
Location: |
Chr.13:33103135-33676835 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD13 | NM_001243476.2 | 18 | 5285 | NP_001230405.1 |
| NM_052851.2 | 14 | 4875 | NP_443083.1 | |
| NM_178006.3 | 14 | 4938 | NP_821074.1 | |
| NM_178007.2 | 14 | 4845 | NP_821075.1 | |
| XM_011535299.2 | 17 | 5031 | XP_011533601.1 | |
| XM_017020834.1 | 14 | 5043 | XP_016876323.1 | |
| XM_017020835.1 | 16 | 4964 | XP_016876324.1 | |
| AL049801.1 | 14 | 4831 | CAB42562.1 | |
| AY082589.1 | 14 | 4933 | AAL91648.1 | |
| AY082590.1 | 14 | 4857 | AAL91649.1 | |
| AY082591.1 | 14 | 4831 | AAL91650.1 | |
| AY366448.1 | AAQ72791.1 | |||
| BX641149.1 | 4 | 1958 | CAE46065.1 | |
| BX647695.1 | 18 | 5245 | CAI46026.1 | |
| CB163673.1 | 1 | 193 |
More Information
Set Membership: |
|

Genomic Map