Assay Details
Target Gene Details
Entrez Gene ID: | 1508 |
Gene Name: | cathepsin B |
Gene Aliases: |
APPS, CPSB |
Location: |
Chr.8:11842524-11868150 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSB | NM_001317237.1 | 10 | 2955 | NP_001304166.1 |
| NM_001908.4 | 10 | 2986 | NP_001899.1 | |
| NM_147780.3 | 12 | 3148 | NP_680090.1 | |
| NM_147781.3 | 11 | 3105 | NP_680091.1 | |
| NM_147782.3 | 11 | 3074 | NP_680092.1 | |
| NM_147783.3 | 11 | 3060 | NP_680093.1 | |
| AK097384.1 | ||||
| AK098296.1 | 1 | 947 | ||
| AK128472.1 | 1 | 3579 | ||
| BC095408.1 | 10 | 2923 | AAH95408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034132 | Chr.8:11799524 - 12340733 on Build GRCh38 | Gain |
|
| esv2762727 | Chr.8:11842150 - 11843744 on Build GRCh38 | Loss |
|
| nsv610293 | Chr.8:11841238 - 12072225 on Build GRCh38 | Gain |
|
| esv34042 | Chr.8:11840828 - 11869308 on Build GRCh38 | Loss |
|
| nsv1018042 | Chr.8:11809987 - 12024876 on Build GRCh38 | Gain |
|
| esv3616259 | Chr.8:11841916 - 11843836 on Build GRCh38 | Loss |
|
| nsv1032739 | Chr.8:11786294 - 11858964 on Build GRCh38 | Loss |
|
| esv2668269 | Chr.8:11841914 - 11843834 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73663020] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map