Assay Details
Target Gene Details
Entrez Gene ID: | 84934 |
Gene Name: | RBPJ interacting and tubulin associated 1 |
Gene Aliases: |
C12orf52, RITA |
Location: |
Chr.12:113185526-113192368 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RITA1 | NM_001286215.1 | 3 | 1298 | NP_001273144.1 |
| NM_032848.2 | 4 | 1430 | NP_116237.1 | |
| XM_005253972.2 | 4 | 1402 | XP_005254029.1 | |
| AK027733.1 | 4 | 1206 | BAB55328.1 | |
| AK027741.1 | 5 | 1206 | BAB55333.1 | |
| AK075358.1 | 4 | 1396 | BAC11568.1 | |
| AK123762.1 | 5 | 1301 | ||
| AK222908.1 | 4 | 1177 | BAD96628.1 | |
| AY007158.1 | 4 | 1218 | ||
| BC022092.2 | 4 | 961 | AAH22092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map