Assay Details
Target Gene Details
Entrez Gene ID: | 7292 |
Gene Name: | tumor necrosis factor superfamily member 4 |
Gene Aliases: |
CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1 |
Location: |
Chr.1:173183729-173462208 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF4 | NM_001297562.1 | 3 | 1565 | NP_001284491.1 |
| NM_003326.4 | 3 | 1607 | NP_003317.1 | |
| XM_011509964.2 | 4 | 2410 | XP_011508266.2 | |
| XM_017002228.1 | 3 | 2839 | XP_016857717.1 | |
| XM_017002229.1 | 5 | 7730 | XP_016857718.1 | |
| XM_017002230.1 | 4 | 2094 | XP_016857719.1 | |
| BC041663.1 | 3 | 1565 | AAH41663.1 | |
| D90224.1 | 3 | 1506 | BAA14259.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116697924] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map