Assay Details
Target Gene Details
Entrez Gene ID: | 134429 |
Gene Name: | StAR related lipid transfer domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.5:111496033-111512597 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STARD4 | NM_001308056.1 | 6 | 1755 | NP_001294985.1 |
| NM_001308059.1 | 7 | 1790 | NP_001294988.1 | |
| NM_001308060.1 | 4 | 1520 | NP_001294989.1 | |
| NM_001308061.1 | 5 | 1647 | NP_001294990.1 | |
| NM_139164.2 | 6 | 1761 | NP_631903.1 | |
| NR_131754.1 | 5 | 1544 | ||
| XM_017009041.1 | 5 | 1572 | XP_016864530.1 | |
| XM_017009042.1 | 5 | 1566 | XP_016864531.1 | |
| XM_017009043.1 | 4 | 1484 | XP_016864532.1 | |
| XM_017009044.1 | 5 | 1554 | XP_016864533.1 | |
| AK054566.1 | 6 | 1623 | BAB70759.1 | |
| AK125317.1 | 5 | 1482 | ||
| AL832599.2 | 2 | 1217 | CAD89942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950113 | Chr.5:111495303 - 111505002 on Build GRCh38 | Duplication |
|
| nsv830449 | Chr.5:111414293 - 111585511 on Build GRCh38 | Gain+Loss |
|
| nsv518727 | Chr.5:111333002 - 111542854 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200864765] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map