Assay Details
Target Gene Details
Entrez Gene ID: | 83695 |
Gene Name: | RAD9-HUS1-RAD1 interacting nuclear orphan 1 |
Gene Aliases: |
C12orf32, HKMT1188, RHINO |
Location: |
Chr.12:2876258-2889525 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHNO1 | NM_001252499.2 | 3 | 1613 | NP_001239428.1 |
| NM_001252500.2 | 3 | 1571 | NP_001239429.1 | |
| NM_001257097.1 | 3 | 1734 | NP_001244026.1 | |
| NM_001257098.1 | 3 | 1633 | NP_001244027.1 | |
| NR_027365.2 | 2 | 1361 | ||
| NR_046432.1 | 4 | 1747 | ||
| NR_046433.1 | 2 | 1535 | ||
| AB073599.1 | 3 | 1589 | BAD38639.1 | |
| AI631627.1 | ||||
| AK021945.1 | 2 | 1337 | ||
| AK304613.1 | 3 | 1547 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain |
|
| esv3628326 | Chr.12:2888954 - 2950527 on Build GRCh38 | Gain |
|
| dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map