Assay Details
Target Gene Details
Entrez Gene ID: | 146956 |
Gene Name: | essential meiotic structure-specific endonuclease 1 |
Gene Aliases: |
MMS4L, SLX2A |
Location: |
Chr.17:50373220-50381479 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EME1 | NM_001166131.1 | NP_001159603.1 | ||
| NM_152463.2 | NP_689676.2 | |||
| XM_005257081.3 | XP_005257138.1 | |||
| XM_011524395.2 | XP_011522697.1 | |||
| XM_017024236.1 | XP_016879725.1 | |||
| AK021607.1 | ||||
| AK055926.1 | BAB71047.1 | |||
| BC016470.2 | AAH16470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833475 | Chr.17:50219520 - 50388833 on Build GRCh38 | Loss |
|
| nsv833476 | Chr.17:50316997 - 50473985 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73989349] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map