Assay Details
Target Gene Details
Entrez Gene ID: | 6391 |
Gene Name: | succinate dehydrogenase complex subunit C |
Gene Aliases: |
CYB560, CYBL, PGL3, QPS1, SDH3 |
Location: |
Chr.1:161314376-161364751 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDHC | NM_001035511.1 | NP_001030588.1 | ||
| NM_001035512.1 | NP_001030589.1 | |||
| NM_001035513.1 | NP_001030590.1 | |||
| NM_001278172.1 | NP_001265101.1 | |||
| NM_003001.3 | NP_002992.1 | |||
| NR_103459.1 | ||||
| AB201252.1 | BAE46977.1 | |||
| AB211234.1 | BAE46978.1 | |||
| AB211235.1 | BAE46979.1 | |||
| AB212048.1 | BAE46980.1 | |||
| AK131051.1 | ||||
| AK293687.1 | ||||
| AK294305.1 | ||||
| BC020808.1 | AAH20808.1 | |||
| BC033626.1 | AAH33626.1 | |||
| BC066329.1 | AAH66329.1 | |||
| BG724095.1 | ||||
| CR457102.1 | CAG33383.1 | |||
| D49737.1 | BAA31998.1 | |||
| DA751501.1 | ||||
| KU178321.1 | ||||
| KU178323.1 | ||||
| KU178324.1 | ||||
| U57877.1 | AAB41838.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010776 | Chr.1:161312594 - 161439428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map