Assay Details
Target Gene Details
Entrez Gene ID: | 25839 |
Gene Name: | component of oligomeric golgi complex 4 |
Gene Aliases: |
CDG2J, COD1 |
Location: |
Chr.16:70480569-70523554 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COG4 | NM_001195139.1 | 18 | 2278 | NP_001182068.1 |
| NM_015386.2 | 19 | 2341 | NP_056201.2 | |
| XM_011522981.2 | 20 | 2501 | XP_011521283.1 | |
| AB088369.1 | 20 | 2477 | BAC05682.1 | |
| AK022874.1 | 19 | 2321 | BAB14286.1 | |
| AK026435.1 | 15 | 1635 | BAB15483.1 | |
| AK096557.1 | 17 | 2059 | BAC04816.1 | |
| AK293235.1 | 16 | 1885 | ||
| AK297557.1 | 20 | 2284 | ||
| AL050101.1 | 15 | 1643 | CAB43272.1 | |
| BC000796.1 | 3 | 339 | AAH00796.1 | |
| BC006306.2 | 16 | 1857 | AAH06306.2 | |
| BC013347.2 | 15 | 1649 | AAH13347.2 | |
| BC027726.2 | 18 | 2252 | AAH27726.1 | |
| BC072438.1 | 19 | 2321 | AAH72438.1 | |
| BX648010.1 | 17 | 3009 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map