Assay Details
Target Gene Details
Entrez Gene ID: | 55626 |
Gene Name: | autophagy and beclin 1 regulator 1 |
Gene Aliases: |
DCAF3, WDR94 |
Location: |
Chr.11:46396412-46594069 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMBRA1 | NM_001267782.1 | 20 | 4805 | NP_001254711.1 |
| NM_001267783.1 | 18 | 4439 | NP_001254712.1 | |
| NM_001300731.1 | 17 | 4616 | NP_001287660.1 | |
| NM_017749.3 | 19 | 4524 | NP_060219.2 | |
| XM_005253009.3 | 18 | 4792 | XP_005253066.1 | |
| XM_005253011.3 | 17 | 4705 | XP_005253068.1 | |
| XM_005253014.3 | 19 | 4522 | XP_005253071.1 | |
| XM_006718259.2 | 18 | 4673 | XP_006718322.1 | |
| XM_006718260.2 | 17 | 4255 | XP_006718323.1 | |
| XM_017018007.1 | 16 | 4525 | XP_016873496.1 | |
| AB051523.1 | 17 | 4568 | ||
| AK000301.1 | 20 | 4755 | BAA91067.1 | |
| AK023197.1 | 12 | 3047 | BAB14457.1 | |
| AK057896.1 | 3 | 1245 | ||
| AK302808.1 | 18 | 4854 | ||
| AL834190.1 | 18 | 4319 | ||
| BC045609.1 | 19 | 4524 | AAH45609.1 | |
| BF589843.1 | ||||
| CA389399.1 | 1 | 109 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626186 | Chr.11:46392885 - 46412875 on Build GRCh38 | Loss |
|
| esv3626185 | Chr.11:46361673 - 46411056 on Build GRCh38 | Loss |
|
| esv3626183 | Chr.11:46227941 - 46507334 on Build GRCh38 | Gain |
|
| nsv309 | Chr.11:46378128 - 46418953 on Build GRCh38 | Deletion |
|
| nsv554202 | Chr.11:46312688 - 46448350 on Build GRCh38 | Loss |
|
| nsv554201 | Chr.11:46260284 - 46401136 on Build GRCh38 | Loss |
|
| nsv825867 | Chr.11:46281633 - 46404063 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map