Assay Details
Target Gene Details
Entrez Gene ID: | 9031 |
Gene Name: | bromodomain adjacent to zinc finger domain 1B |
Gene Aliases: |
WBSCR10, WBSCR9, WSTF |
Location: |
Chr.7:73440398-73522298 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAZ1B | NM_032408.3 | NP_115784.1 | ||
| XM_017012773.1 | XP_016868262.1 | |||
| AB032253.1 | BAA89210.1 | |||
| AF072810.1 | AAC97879.1 | |||
| AF084479.1 | AAD08675.1 | |||
| AK123274.1 | 1 | 777 | ||
| BC136520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759537 | Chr.7:72446485 - 73461568 on Build GRCh38 | Gain+Loss |
|
| dgv6467n100 | Chr.7:73420075 - 73583448 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map