Assay Details
Target Gene Details
Entrez Gene ID: | 84894 |
Gene Name: | leucine rich repeat and Ig domain containing 1 |
Gene Aliases: |
LERN1, LRRN6A, UNQ201 |
Location: |
Chr.15:77613024-77820900 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINGO1 | NM_001301186.1 | 6 | 2666 | NP_001288115.1 |
| NM_001301187.1 | 6 | 2683 | NP_001288116.1 | |
| NM_001301189.1 | 6 | 2677 | NP_001288118.1 | |
| NM_001301191.1 | 6 | 2862 | NP_001288120.1 | |
| NM_001301192.1 | 5 | 2800 | NP_001288121.1 | |
| NM_001301194.1 | 4 | 2618 | NP_001288123.1 | |
| NM_001301195.1 | 4 | 2553 | NP_001288124.1 | |
| NM_001301197.1 | 4 | 2566 | NP_001288126.1 | |
| NM_001301198.1 | 3 | 2363 | NP_001288127.1 | |
| NM_001301199.1 | 4 | 2536 | NP_001288128.1 | |
| NM_001301200.1 | 4 | 2609 | NP_001288129.1 | |
| NM_032808.6 | 2 | 2381 | NP_116197.4 | |
| XM_011522118.2 | 2 | 2293 | XP_011520420.1 | |
| XM_017022682.1 | 7 | 3009 | XP_016878171.1 | |
| AK027500.1 | 1 | 1314 | BAB55157.1 | |
| AL109677.1 | ||||
| AL834260.1 | 1 | 2041 | CAD38935.1 | |
| AY324320.1 | 2 | 2221 | AAQ97216.1 | |
| AY324322.1 | AAQ97217.1 | |||
| AY358284.1 | AAQ88651.1 | |||
| BC011057.1 | AAH11057.2 | |||
| BC068558.1 | AAH68558.1 | |||
| H11343.1 | ||||
| HY342106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833060 | Chr.15:77524312 - 77742862 on Build GRCh38 | Gain |
|
| nsv1036749 | Chr.15:77591684 - 77620066 on Build GRCh38 | Gain |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map