Assay Details
Target Gene Details
Entrez Gene ID: | 8828 |
Gene Name: | neuropilin 2 |
Gene Aliases: |
NP2, NPN2, PRO2714, VEGF165R2 |
Location: |
Chr.2:205682450-205798133 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRP2 | NM_003872.2 | 1 | 372 | NP_003863.2 |
| NM_018534.3 | 1 | 372 | NP_061004.3 | |
| NM_201264.1 | 1 | 372 | NP_957716.1 | |
| NM_201266.1 | 1 | 372 | NP_957718.1 | |
| NM_201267.1 | 1 | 372 | NP_957719.1 | |
| NM_201279.1 | 1 | 372 | NP_958436.1 | |
| XM_005246933.3 | XP_005246990.2 | |||
| XM_005246934.3 | XP_005246991.2 | |||
| XM_017005185.1 | XP_016860674.1 | |||
| XM_017005186.1 | XP_016860675.1 | |||
| XM_017005187.1 | 1 | 422 | XP_016860676.1 | |
| XM_017005188.1 | 1 | 421 | XP_016860677.1 | |
| AK130198.1 | 2 | 358 | ||
| AK290934.1 | ||||
| AL833606.1 | 1 | 372 | ||
| BC009222.2 | AAH09222.1 | |||
| BX537423.1 | 1 | 373 | CAD97665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834514 | Chr.2:205575978 - 205738443 on Build GRCh38 | Gain |
|
| nsv3123 | Chr.2:205681544 - 205714569 on Build GRCh38 | Insertion |
|
| esv25775 | Chr.2:205407368 - 205806595 on Build GRCh38 | Gain |
|
| nsv584246 | Chr.2:205627674 - 205747861 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map