Assay Details
Target Gene Details
Entrez Gene ID: | 344892 |
Gene Name: | receptor transporter protein 2 |
Gene Aliases: |
Z3CXXC2 |
Location: |
Chr.3:187698259-187715625 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTP2 | NM_001004312.2 | 2 | 1044 | NP_001004312.2 |
| XM_017006301.1 | 5 | 1238 | XP_016861790.1 | |
| XM_017006302.1 | 5 | 1189 | XP_016861791.1 | |
| AY562236.1 | 2 | 615 | AAT70681.1 | |
| BC068081.1 | 2 | 1044 | AAH68081.1 |
More Information
Set Membership: |
|

Genomic Map