Assay Details
Target Gene Details
Entrez Gene ID: | 2138 |
Gene Name: | EYA transcriptional coactivator and phosphatase 1 |
Gene Aliases: |
BOP, BOR, BOS1, OFC1 |
Location: |
Chr.8:71197433-71548130 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA1 | NM_000503.5 | 18 | 3478 | NP_000494.2 |
| NM_001288574.1 | 18 | 3460 | NP_001275503.1 | |
| NM_001288575.1 | 18 | 3396 | NP_001275504.1 | |
| NM_172058.3 | 17 | 3070 | NP_742055.1 | |
| NM_172059.3 | 16 | 2787 | NP_742056.1 | |
| NM_172060.3 | 16 | 2942 | NP_742057.1 | |
| XM_011517483.2 | 17 | 7775 | XP_011515785.1 | |
| XM_011517484.2 | 19 | 3221 | XP_011515786.2 | |
| XM_017013201.1 | 19 | 3264 | XP_016868690.1 | |
| XM_017013202.1 | 19 | 3108 | XP_016868691.1 | |
| XM_017013203.1 | 19 | 3233 | XP_016868692.1 | |
| XM_017013204.1 | 19 | 3219 | XP_016868693.1 | |
| XM_017013206.1 | 20 | 3217 | XP_016868695.1 | |
| XM_017013207.1 | 18 | 3145 | XP_016868696.1 | |
| XM_017013208.1 | 18 | 3500 | XP_016868697.1 | |
| XM_017013209.1 | 18 | 3020 | XP_016868698.1 | |
| XM_017013210.1 | 18 | 3129 | XP_016868699.1 | |
| XM_017013211.1 | 18 | 3108 | XP_016868700.1 | |
| XM_017013212.1 | 17 | 3375 | XP_016868701.1 | |
| XM_017013213.1 | 17 | 7913 | XP_016868702.1 | |
| AJ000097.1 | 17 | 3015 | CAA03922.1 | |
| AJ000098.1 | 18 | 3476 | CAA03923.1 | |
| AK127798.1 | 18 | 2896 | ||
| BQ223588.1 | 1 | 438 |
More Information
Set Membership: |
|

Genomic Map