Assay Details
Target Gene Details
Entrez Gene ID: | 149428 |
Gene Name: | BCL2 interacting protein like |
Gene Aliases: |
BNIP-S, BNIP-Salpha, BNIP-Sbeta, BNIPL-1, BNIPL-2, BNIPL1, BNIPL2, BNIPS, PP753 |
Location: |
Chr.1:151036553-151047600 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BNIPL | NM_001159642.1 | 10 | 1554 | NP_001153114.1 |
| NM_138278.3 | 10 | 1302 | NP_612122.2 | |
| XM_011509236.1 | 10 | 1255 | XP_011507538.1 | |
| XM_017000420.1 | 11 | 1349 | XP_016855909.1 | |
| AF193056.1 | 10 | 1500 | AAG22484.1 | |
| AK092595.1 | 10 | 1254 | ||
| AY033000.1 | 10 | 1286 | AAK54348.1 | |
| BC027868.1 | 10 | 1279 | ||
| BC074779.2 | AAH74779.3 | |||
| BC074780.2 | AAH74780.3 | |||
| CU451236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map