Assay Details
Target Gene Details
Entrez Gene ID: | 102465667 |
Gene Name: | microRNA 7110 |
Gene Aliases: |
hsa-mir-7110 |
Location: |
Chr.3:123161794-123161879 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR7110 | NR_106960.1 |
Target Gene Details
Entrez Gene ID: | 10954 |
Gene Name: | protein disulfide isomerase family A member 5 |
Gene Aliases: |
PDIR |
Location: |
Chr.3:123067009-123162106 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIA5 | NM_006810.3 | NP_006801.1 | ||
| NR_028444.1 | ||||
| AK054963.1 | 5 | 2058 | ||
| BC001625.1 | AAH01625.1 | |||
| CA414869.1 | ||||
| CB158962.1 | ||||
| D49490.1 | BAA08451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521590 | Chr.3:123161344 - 123169790 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map