Assay Details
Target Gene Details
Entrez Gene ID: | 55687 |
Gene Name: | tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase |
Gene Aliases: |
LCAL3, MTO2, MTU1, TRMT, TRMT1, TRNT1 |
Location: |
Chr.22:46335401-46357340 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMU | NM_001282782.1 | NP_001269711.1 | ||
| NM_001282783.1 | NP_001269712.1 | |||
| NM_001282784.1 | NP_001269713.1 | |||
| NM_001282785.1 | NP_001269714.1 | |||
| NM_018006.4 | NP_060476.2 | |||
| NR_104240.1 | ||||
| NR_104241.1 | ||||
| XM_005261678.1 | XP_005261735.1 | |||
| XM_005261681.1 | XP_005261738.1 | |||
| XM_011530271.2 | XP_011528573.1 | |||
| XM_011530272.1 | XP_011528574.1 | |||
| XM_011530273.1 | XP_011528575.1 | |||
| XM_011530274.2 | XP_011528576.1 | |||
| XM_011530275.1 | XP_011528577.1 | |||
| AB178028.1 | BAD66875.1 | |||
| AK000212.1 | 1 | 1293 | ||
| AK001002.1 | BAA91462.1 | |||
| AK055295.1 | 2 | 1598 | ||
| AK093743.1 | ||||
| AK290712.1 | ||||
| AY062123.2 | AAL38183.1 | |||
| BC027991.1 | ||||
| CR456445.1 | CAG30331.1 | |||
| DQ309998.1 | ABC46690.1 | |||
| DQ309999.1 | ABC46691.1 | |||
| DQ310000.1 | ABC46692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955177 | Chr.22:46350404 - 46389303 on Build GRCh38 | Deletion |
|
| nsv829293 | Chr.22:46241054 - 46741639 on Build GRCh38 | Gain |
|
| nsv829281 | Chr.22:45861485 - 46735858 on Build GRCh38 | Gain |
|
| nsv834225 | Chr.22:46323584 - 46510799 on Build GRCh38 | Loss |
|
| nsv471214 | Chr.22:46278315 - 46367774 on Build GRCh38 | Loss |
|
| nsv1066865 | Chr.22:46352413 - 46490139 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map