Assay Details
Target Gene Details
Entrez Gene ID: | 27345 |
Gene Name: | potassium calcium-activated channel subfamily M regulatory beta subunit 4 |
Gene Aliases: |
- |
Location: |
Chr.12:70366220-70434292 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNMB4 | NM_014505.5 | 1 | 638 | NP_055320.4 |
| XM_011538188.2 | 1 | 700 | XP_011536490.1 | |
| AF160967.1 | 1 | 522 | AAF69805.1 | |
| AF207992.1 | 1 | 185 | AAF28333.1 | |
| AF215891.1 | 1 | 240 | AAF75596.1 | |
| BC042446.2 | 1 | 347 | AAH42446.2 | |
| BC050621.1 | 1 | 279 | AAH50621.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559257 | Chr.12:70334144 - 70377881 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map