Assay Details
Target Gene Details
Entrez Gene ID: | 11047 |
Gene Name: | adhesion regulating molecule 1 |
Gene Aliases: |
ARM-1, ARM1, GP110 |
Location: |
Chr.20:62302056-62308862 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRM1 | NM_001281437.1 | NP_001268366.1 | ||
| NM_001281438.1 | NP_001268367.1 | |||
| NM_007002.3 | NP_008933.2 | |||
| NM_175573.2 | NP_783163.1 | |||
| XM_005260257.1 | XP_005260314.1 | |||
| XM_011528503.1 | XP_011526805.1 | |||
| XM_017027602.1 | XP_016883091.1 | |||
| XM_017027603.1 | XP_016883092.1 | |||
| XM_017027604.1 | XP_016883093.1 | |||
| XM_017027605.1 | XP_016883094.1 | |||
| XM_017027606.1 | XP_016883095.1 | |||
| XM_017027607.1 | XP_016883096.1 | |||
| XM_017027608.1 | XP_016883097.1 | |||
| XM_017027609.1 | XP_016883098.1 | |||
| XM_017027610.1 | XP_016883099.1 | |||
| AK297567.1 | ||||
| AL558522.3 | ||||
| BC010733.1 | AAH10733.1 | |||
| BC017245.2 | AAH17245.1 | |||
| BG337785.1 | ||||
| CR749213.1 | 1 | 2746 | CAH18070.2 | |
| D64154.1 | BAA11023.1 | |||
| HY178253.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication |
|
| nsv953301 | Chr.20:62305045 - 62338544 on Build GRCh38 | Deletion |
|
| nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss |
|
| nsv3441 | Chr.20:62297903 - 62320059 on Build GRCh38 | Insertion |
|
| nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss |
|
| nsv834024 | Chr.20:62259914 - 62455522 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map